Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33125-U

Supelco

BF3 - Butanol solution

10 % (w/w)

Synonym(s):

Boron trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
23151816
PubChem Substance ID:

grade

derivatization grade ((GC derivatization))

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 1 × 100 mL (33125-U)

concentration

10 % (w/w)

SMILES string

FB(F)F

InChI

1S/BF3/c2-1(3)4

InChI key

WTEOIRVLGSZEPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Boron trifluoride is a colourless gas which readily reacts with water to form hydrates. It has been widely used as catalyst for polymerizations, alkylations and condensation reactions. It is used as gas flux for internal soldering or brazing. It also finds application in being used as extinguisher for magnesium fires. Furthermore esterification by BF3-butanol helps in obtaining volatile derivatives of aliphatic acids such as hydroxyacids.

Application

It was used in extraction and derivatization of γ−hydroxybutyrate from human plasma or urine during a study to determine 1,4-butanediol and γ-hydroxybutyrate using capillary gas chromatography.

Other Notes

Reagent for butyl esters and trifluoroacetyl.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

99.0 °F - closed cup

flash_point_c

37.2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J L Iverson et al.
Journal - Association of Official Analytical Chemists, 60(2), 284-288 (1977-03-01)
Butter oil is extracted from butter with hexane, using a rolling boil technique. The resulting 10% butter oil in hexane is converted to butyl esters, using butanol in place of methanol as specified in the official final action method, 28.063-28.067.
M.A. Lambert and C.W. Moss
Journal of Chromatography A, 74, 335-335 (1972)
Compressed Gas Association
Handbook of Compressed Gases, 271-271 (2012)
Panayiotis V Ioannou et al.
Chemistry and physics of lipids, 163(1), 51-55 (2009-11-11)
The crucial step in the preparation of the title arsonolipids starting from the dichloromethane-soluble dithioarsonite CH(2)(OH)CH(OH)CH(2)-As(SPh)(2) is to avoid an internal cyclization during the acylation which protects the primary -OH group from being acylated. This was to a large extent
David Chivall et al.
Rapid communications in mass spectrometry : RCM, 26(10), 1232-1240 (2012-04-14)
Compound-specific stable hydrogen isotope analysis of fatty acids is being used increasingly as a means of deriving information from a diverse range of materials of archaeological, geological and environmental interest. Preparative steps required prior to isotope ratio mass spectrometry (IRMS)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service