Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33084

Supelco

BSTFA, Derivatization Grade

derivatization grade (GC derivatization)

Synonym(s):

N,O-Bis(trimethylsilyl)trifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

derivatization grade (GC derivatization)

description

Silylation reagent

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 144 × 0.1 mL

technique(s)

GC/MS: suitable

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3

InChI key

XCOBLONWWXQEBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of acetaminophen, acetanilide, n-acetylneuraminic acid, U-aminobutyric acid, arginine, dipeptides, dopamine O-sulfate (3- and 4-isomers), glycine, amino sugar methyl glycosides, glycosphingolipi, n-hydroxylamines, lysine, nitrilotriacetate, phenacetin, phenols, phospholipids, phosphoserine and phosphothreonine, prostaglandin H2 methyl ester, sulfur amino acids and terephthalate prepolymer mixture, and thromboxane B2.

Other Notes

Reagent for N-acetyl, O-trimethylesilyl, trimethylsilyl, trimethylsilyl diglyceride, sulfonyl trimethylsilyl esters, thiohydantoins of amino acids., and trimethylsilyl thiohydantoins.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

93.2 °F

flash_point_c

34 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E.R.Atkinson and S.I. Calouche
Analytical Chemistry, 43, 460-460 (1971)
C.W. Gehrke and K. Leimer
Journal of Chromatography A, 57, 219-219 (1971)
B. Wengle and K.Hellstrom
Scandinavian Journal of Clinical and Laboratory Investigation, 28, 477-477 (1977)
C.W. Gehrke and K. Leimer
Journal of Chromatography A, 53, 201-201 (1970)
Rapid determination of twenty amino acids by gas chromatography.
J P Hardy et al.
Analytical chemistry, 44(8), 1497-1499 (1972-07-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service