Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33080

Supelco

Methanolic Base

0.5 M CH3ONa (0.5N), for GC derivatization

Synonym(s):

Sodium methoxide solution, Sodium methylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3ONa
CAS Number:
Molecular Weight:
54.02
Beilstein/REAXYS Number:
3592982
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:

grade

derivatization grade ((GC derivatization))
for GC derivatization

packaging

pkg of 100 mL

concentration

0.5 M CH3ONa (0.5N)

storage temp.

2-8°C

SMILES string

[Na+].C[O-]

InChI

1S/CH3O.Na/c1-2;/h1H3;/q-1;+1

InChI key

WQDUMFSSJAZKTM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 1

target_organs

Eyes,Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

53.6 °F - closed cup

flash_point_c

12 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hormoz Azizian et al.
Lipids, 40(8), 855-867 (2005-11-22)
A rapid method was developed for classifying and quantifying the FA composition of edible oils and fats using Fourier Transform near infrared spectroscopy (FT-NIR). The FT-NIR spectra showed unique fingerprints for saturated FA, cis and trans monounsaturated FA, and all
P. D. Chaithanya Sudha
Pharmaceutical Analysis, 88-88 (2013)
Dauenpen Meesapyodsuk et al.
Journal of lipid research, 56(11), 2102-2109 (2015-09-02)
Ricinoleic acid (12-hydroxyoctadec-cis-9-enoic acid) has many specialized uses in bioproduct industries, while castor bean is currently the only commercial source for the fatty acid. This report describes metabolic engineering of a microbial system (Pichia pastoris) to produce ricinoleic acid using
Gema Flores et al.
Food chemistry, 194, 1260-1265 (2015-10-17)
Black currant seeds are obtained as a residue during juice production. Black currant seed oil contains high amounts of nutritionally desirable constituents such as γ-linolenic acid (GLA), α-linolenic acid (ALA) and stearidonic acid (SA), as well as certain phenolic acids
Holger Pletsch et al.
Nanoscale, 7(5), 1977-1983 (2014-12-24)
The design of networks from polymers and noble metal nanoparticles requires thorough control over topological polymer-particle arrangements. This study explores the interaction between a linear polydentate poly(propylene sulfide) (PPrS) ligand and silver nanoparticles (AgNPs) with an aim to study its

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service