Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33047-U

Supelco

Phospray

pkg of 200 mL

Synonym(s):

Supelco® TLC Spray Reagents, Phospray, 200ml

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

description

Phospray

usage

For the detection of phospholipids

packaging

pkg of 200 mL

Looking for similar products? Visit Product Comparison Guide

Legal Information

Supelco is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

signalword

Danger

Hazard Classifications

Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Gavilanes et al.
The Journal of biological chemistry, 257(13), 7770-7777 (1982-07-10)
The lipid composition of hepatitis B surface antigen (HBsAg) (subtype adw) obtained from different carriers has been determined and proven to be truly characteristic of HBsAg and not subject to individual variation. Phosphatidylcholine (approximately 60%), cholesteryl ester (approximately 14%), cholesterol
D G Cooper et al.
Applied and environmental microbiology, 42(3), 408-412 (1981-09-01)
The lipopeptide, surfactin, is produced by Bacillus subtilis. A study has been made on large-scale production of this surfactant. A good yield was obtained from a glucose substrate fermentation by continuously removing the product by foam fractionation. The surfactin could
Gregor Cevc
Phospholipids Handbook, 146-146 (1993)
E S Kaneshiro et al.
Journal of lipid research, 27(12), 1294-1303 (1986-12-01)
Inositol lipids account for 15% of the total cellular phospholipids of Leishmania donovani promastigotes. Four major inositol lipids were identified and characterized: phosphatidylinositol (PI), phosphatidylinositol phosphate (PI-P), phosphatidylinositol diphosphate (PI-P2), and an inositol sphingophospholipid (InSL). Diacyl and alkyl acyl PI

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service