Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33035-U

Supelco

BSA Derivatization Grade

derivatization grade, Silylation reagent

Synonym(s):

N,O-Bis(trimethylsilyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151816

grade

derivatization grade ((GC derivatization))
derivatization grade

reaction suitability

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 144 × 0.1 mL (33035-U)

Looking for similar products? Visit Product Comparison Guide

General description

BSA (N,O-Bis(trimethylsilyl)acetamide) is a powerful reagent for silyation of alkyl hydroperoxides.[1] It reacts quantitatively under mild conditions with alcohols, amines, phenols and carboxylic acids which gives trimethylsilyl ethers and esters.[2]

Application

BSA was used in silyation procedure of amino acid phenylthiohydantoin standards during GC analysis.[3]

Other Notes

Reagent for 1,3-dipheylimidazolidine-2-carboxylic acid, o-ethyloxime, quinoxalone, trimethylsilyl and trimethylsilyl oximes.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 3 - Skin Corr. 1B

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

111.2 °F - closed cup

flash_point_c

44 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Knapp
Handbook of Analytical Derivatization Reactions, 202-214 (1979)
Gas chromatographic behavior of trimethylsilylated phenylthiohydantoin amino acids.
R E Harman et al.
Analytical biochemistry, 25(1), 452-458 (1968-10-24)
Mass spectrometric studies of thyroxine and related compounds. Trimethylsilyl derivatives.
A M Lawson et al.
Biomedical mass spectrometry, 1(6), 374-380 (1974-12-01)
The quantitative extraction and gas-liquid chromatographic determination of organic acids in urine.
R A Chalmers et al.
The Analyst, 97(161), 958-967 (1972-12-01)
S.Mori et al.
Analytical Chemistry, 42, 959-959 (1970)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service