Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33033

Supelco

Boron trichloride - Methanol

12 % (w/w), pkg of 400 mL

Synonym(s):

BCl3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116105

reaction suitability

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 400 mL

concentration

12 % (w/w)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Boron trichloride-methanol is a derivatizing reagent. It is basically used in the esterification of fatty acids to fatty acid methyl ester (FAME).

Application

Boron trichloride-Methanol may be used in converting linoleate to methyl linoleate for understanding the effect of palmitic acid on oxidation of oleic acid.[1]

Other Notes

Reagent for methyl ester, pentafluoropropionyl.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Repr. 1B - Skin Corr. 1B - STOT SE 1

target_organs

Eyes,Central nervous system

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

51.8 °F - closed cup

flash_point_c

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J.Segura et al.
Biomedical Mass Spectrometry, 3, 91-91 (1971)
D.R. Knapp
Handbook of Analytical Derivatization Reactions, 218-218 (1979)
J M Onorato et al.
The Journal of biological chemistry, 275(28), 21177-21184 (2000-05-10)
Maillard or browning reactions lead to formation of advanced glycation end products (AGEs) on protein and contribute to the increase in chemical modification of proteins during aging and in diabetes. AGE inhibitors such as aminoguanidine and pyridoxamine (PM) have proven
Gas chromatography-mass spectrometry of catecholamines and tryptamines: determination of gas chromatographic profiles of the amines, their precursors and their metabolites.
E G Peralta et al.
Journal of chromatographic science, 12(11), 701-709 (1974-11-01)
Chanhee Jo et al.
Experimental & molecular medicine, 52(12), 2005-2019 (2020-12-15)
Acetylation is the most studied histone acyl modification and has been recognized as a fundamental player in metabolic gene regulation, whereas other short-chain acyl modifications have only been recently identified, and little is known about their dynamics or molecular functions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service