Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33024

Supelco

BSTFA, Derivatization Grade

derivatization grade (GC derivatization)

Synonym(s):

N,O-Bis(trimethylsilyl)trifluoroacetamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

derivatization grade (GC derivatization)

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 20 × 1 mL

technique(s)

GC/MS: suitable

InChI

1S/C8H18F3NOSi2/c1-14(2,3)12-7(8(9,10)11)13-15(4,5)6/h1-6H3

InChI key

XCOBLONWWXQEBS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for the derivatization of acetaminophen, acetanilide, n-acetylneuraminic acid, U-aminobutyric acid, sulfur amino acids, arginine, dipeptides, dopamine O-sulfate (3- and 4-isomers), amino sugar methyl glycosides, glycosphingolipi, glysine, n-hydroxylamines, lysine, nitrilotriacetate, phenacetin, phenols, phenols (sterically hindered), phospholipids, phosphoserine and phosphothreonine, prostaglandins F2α, prostaglandin H2 methyl ester, psiocin, psiocybin, dopamine O-sulfate (3- and 4-isomers), terephthalate prepolymer mixture, thiohydantoins of amino acids, and thromboxane B2.

Other Notes

Reagent for N-acetyl, O-trimethylesilyl, trimethylsilyl, trimethylsilyl ether, trimethylsilyl thiohydantoins, sulfonyl trimethylsilyl esters and trimethylsilyl diglyceride.

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

93.2 °F

flash_point_c

34 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C.W. Gehrke and K. Leimer
Journal of Chromatography A, 59, 219-219 (1971)
E.R.Atkinson and S.I. Calouche
Analytical Chemistry, 43, 460-460 (1971)
D B Repke et al.
Journal of pharmaceutical sciences, 66(5), 743-744 (1977-05-01)
With the combined technique of GLC-mass spectrometry, psilocin and psilocybin, two hallucinogenic indoles, were analyzed as their trimethylsilyl derivatives. The method was applied to these two components in an extract of Psilocybe cubensis (Earle) Sing.
A new silylation reagent for amino acids bis(trimethylsilyl)trifluoroacetamide (BSTFA).
D L Stalling et al.
Biochemical and biophysical research communications, 31(4), 616-622 (1968-05-23)
N.E. Hoffman and K.A. Peteranetz
Analytical Letters, 5, 589-589 (1972)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service