Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33020-U

Supelco

BF3 - Methanol

10 % (w/w)

Synonym(s):

Boron trifluoride

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

grade

derivatization grade ((GC derivatization))
for GC derivatization

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

reagent type: derivatization reagent
reaction type: Esterifications

packaging

pkg of 19 × 2 mL (33020-U)

concentration

10 % (w/w)

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Boron trifluoride is a colourless gas and readily reacts with water to form hydrates. It has been widely used as a catalyst during polymerizations, alkylations and condensation reactions. It is generally used as gas flux during internal soldering or brazing. It also finds application in being used a extinguisher for magnesium fires.[1]

Application

It was used in transmethylation of 1,2-Diacylglycerol (DAG) containing fractions which were purified using HPLC.[2] It may also be used to treat lipids during determination of fatty acid methyl esters and dimethylacetals using GC.[3]

Other Notes

Reagent for O-acetyl, methyl ester, methyl 2,2-dimethoxypropionate, dimethyl acetals and dimethoxy.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 1

target_organs

Eyes, Kidney

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

60.8 °F - closed cup

flash_point_c

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S B Bocckino et al.
The Journal of biological chemistry, 260(26), 14201-14207 (1985-11-15)
1,2-Diacylglycerol (DAG) was measured in neutral lipid extracts from isolated hepatocytes using high pressure liquid chromatography followed by refractive index detection. Maximally effective doses of epinephrine, angiotensin II, and vasopressin increased DAG by approximately 65, 80, and 180-250%, respectively, with
PREPARATION OF FATTY ACID METHYL ESTERS AND DIMETHYLACETALS FROM LIPIDS WITH BORON FLUORIDE--METHANOL.
W R MORRISON et al.
Journal of lipid research, 5, 600-608 (1964-10-01)
Florence R Danila et al.
Communications biology, 4(1), 254-254 (2021-02-28)
C4 photosynthesis provides an effective solution for overcoming the catalytic inefficiency of Rubisco. The pathway is characterised by a biochemical CO2 concentrating mechanism that operates across mesophyll and bundle sheath (BS) cells and relies on a gas tight BS compartment.
Compressed Gas Association
Handbook of Compressed Gases, 271-271 (2012)
ANALYSIS OF HYDROXY FATTY ACIDS BY GAS-LIQUID CHROMATOGRAPHY.
J S O'BRIEN et al.
Analytical biochemistry, 7, 288-296 (1964-03-01)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service