Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

33014

Supelco

Chlorotrimethylsilane

pkg of 100 mL

Synonym(s):

TMCS, Derivatization Grade, TMCS, Trimethylchlorosilane, Trimethylsilyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116105

grade

derivatization grade ((GC derivatization))
derivatization grade

description

Silylation reagent

form

liquid

reaction suitability

reagent type: derivatization reagent
reaction type: Acylations

reagent type: derivatization reagent
reaction type: Silylations

packaging

pkg of 100 mL

technique(s)

GC/MS: suitable

General description

Chlorotrimethylsilane is widely used in characterising organolithium compounds. It is also used in establishing the degree of polymetallation by organolithium compounds.[1]

Application

It was used to reconstitute the phytic acid residue during derivatization procedure, before injecting the residue into GC for determination of phytic acid content in biological samples.[2] It was also used in toluene for silanizing all the glassware used for detecting, identifying and quantifying polychlorinated biphenyls (PCB) using solid-phase microextraction combined with gas chromatography with electron capture detection.[3]

Other Notes

Reagent for trimethylsilyl chlorohydrin, trimethylsilyl and trimethylsilyl ether.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A

supp_hazards

Storage Class

3 - Flammable liquids

wgk_germany

WGK 1

flash_point_f

-18.4 °F - closed cup

flash_point_c

-28 °C - closed cup

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Knapp
Handbook of Analytical Derivatization Reactions, 472-472 (1979)
Cláudia H Kowalski et al.
Talanta, 116, 315-321 (2013-10-24)
Due to the tendency of polychlorinated biphenyls (PCB) to accumulate in matrixes with high lipid content, the contamination of the breast milk with these compounds is a serious issue, mainly to the newborn. In this study, milk samples were collected
Jszafranek et al.
Journal of Chromatography A, 88, 149-149 (1974)
B. J. Wakefield
The Chemistry of Organolithium Compounds, 260-260 (2013)
M.G.Horning et al.
Journal of Gas Chromatography, 5, 297-297 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service