Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300500

Oxygen

99.6%

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12141904
Pricing and availability is not currently available.

size

Scotty 14

assay

99.6%

form

solid

packaging

pkg of 14 L

manufacturer/tradename

Scott Gas


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pu Yang et al.
Tissue engineering. Part A, 26(19-20), 1112-1122 (2020-04-24)
A central challenge in tissue engineering is obtaining a suitable cell type with a capable delivery vehicle to replace or repair damaged or diseased tissues with tissue mimics. Notably, for skeletal muscle tissue engineering, given the inadequate availability and regenerative
Jae Soon Kang et al.
PloS one, 6(4), e19063-e19063 (2011-05-11)
The pinewood nematode genome encodes at least three distinct acetylcholinesterases (AChEs). To understand physiological roles of the three pinewood nematode AChEs (BxACE-1, BxACE-2, and BxACE-3), BxACE-3 in particular, their tissue distribution and inhibition profiles were investigated. Immunohistochemistry revealed that BxACE-1
Scott W Eastman et al.
The Journal of cell biology, 184(6), 881-894 (2009-03-25)
HECT (homologous to the E6AP C terminus) ubiquitin ligases have diverse functions in eukaryotic cells. In screens for proteins that bind to the HECT ubiquitin ligase WWP1, we identified Spartin, which is also known as SPG20. This protein is truncated
Esther Asedegbega-Nieto et al.
Nanoscale research letters, 6(1), 353-353 (2011-06-30)
Carbon nanotubes [CNTs] were synthesized by the catalytic vapor decomposition method. Thereafter, they were functionalized in order to incorporate the oxygen groups (OCNT) and subsequently the amine groups (ACNT). All three CNTs (the as-synthesized and functionalized) underwent reaction with an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service