Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300100

Supelco

Hydrogen (99.99%)

cylinder of 14 L, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
23151816

grade

analytical standard

assay

99.99%

form

gas

packaging

cylinder of 14 L

manufacturer/tradename

Scott Gas T108-14

availability

not available in EU, Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

format

multi-component solution

General description

Specifications:
Contents
: 14 liters
Pressure: 240 psig
Outlet fitting: CGA-160-1/8 inch, NPT F
Weight: 1.5 lb.
Dimensions
: 3 x 11 inches
D.O.T. specifications: 4B240

Application

Hydrogen (99.99%) along with gas calibration standard has been used to check the gas storage capability of Monoject tubes during post-mortem examination of seals and dolphins.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Features and Benefits

  • Hydrogen (99.99%) gas mixture is suitable for GC calibration, testing of fixed and personal gas monitors and in situ industrial stack analyzers.
  • A certificate of analysis is included.
  • Packaged in lightweight, non-refillable, recyclable containers.
  • Portable, convenient, and easy to use.
  • Manufactured gravimetrically for maximum accuracy.
  • NIST traceable.

pictograms

FlameGas cylinder

signalword

Danger

hcodes

Hazard Classifications

Flam. Gas 1A - Press. Gas Compr. Gas

Storage Class

2A - Gases

wgk_germany

nwg

flash_point_f

<-238.0 °F - closed cup

flash_point_c

< -150 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Compositional discrimination of decompression and decomposition gas bubbles in bycaught seals and dolphins.
Bernaldo de Quiros Y
PLoS ONE, 8(12), e83994-e83994 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service