Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29749-U

Supelco

Tenax® GR

60/80 mesh, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, sealed with Brass Endcaps, preconditioned, pkg of 10 ea

Synonym(s):

Tenax® GR

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

material

glass TD tube

feature

fritted
preconditioned

packaging

pkg of 10 ea

technique(s)

active air sampling: suitable

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Tenax GR graphitized porous polymer

particle size

60/80 mesh

suitability

suitable for (extends range of Tenax TA)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with PerkinElmer, Markes, DANI, OI Analytical, and Shimadzu Instruments

Looking for similar products? Visit Product Comparison Guide

Legal Information

Tenax is a registered trademark of Buchem B.V.

Pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service