Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29553-U

Supelco

High Capacity Gas Purifier, Replacement Heating Element

AC/DC input 110 V AC, pack of 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121813
NACRES:
SB.54

packaging

pack of 2

Looking for similar products? Visit Product Comparison Guide

General description

This replacement element can be used to replace either the top or bottom half of a clam-shell oven.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service