29547-U
High Capacity Gas Purifier, Starter Kit
AC/DC input 230 V AC, size 1/4 in., CE Approved
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
SB.54
size
1/4 in.
Looking for similar products? Visit Product Comparison Guide
General description
Conformity Marks: CE Approved
No other purifier removes both moisture and oxygen in such large quantities (35 liters of moisture, 14 liters of oxygen ).
Protects GC columns and detectors from damage caused by moisture and oxygen in the carrier gas stream, even when present up to 100 ppm levels. The clam-shell oven heats a converter tube to 580°C, causing moisture and oxygen to irreversibly react with the reactant material. This chemical reaction prevents contaminants from returning to the gas stream, even when the material approaches saturation, or when the oven is cooled. Carbon monoxide and carbon dioxide are also removed.
It is recommended to install 4 ft. (1.2 m) of tubing downstream of the oven to allow heat to dissipate from the gas stream. This tubing can be coiled to reduce space.
The starter kit includes a clam-shell oven, two elements (installed), and a converter tube.
Note: Not for use with hydrogen gas.
Clam-Shell Oven:
Converter Tube:
Protects GC columns and detectors from damage caused by moisture and oxygen in the carrier gas stream, even when present up to 100 ppm levels. The clam-shell oven heats a converter tube to 580°C, causing moisture and oxygen to irreversibly react with the reactant material. This chemical reaction prevents contaminants from returning to the gas stream, even when the material approaches saturation, or when the oven is cooled. Carbon monoxide and carbon dioxide are also removed.
It is recommended to install 4 ft. (1.2 m) of tubing downstream of the oven to allow heat to dissipate from the gas stream. This tubing can be coiled to reduce space.
The starter kit includes a clam-shell oven, two elements (installed), and a converter tube.
Note: Not for use with hydrogen gas.
Clam-Shell Oven:
- Length: 14 1/2 in. (36.8 cm)
- Height: 5 1/4 in. (13.3 cm)
- Depth: 6 in. (15.2 cm)
- Operating Temp.: 580°C
- Power Consumption: 90 watt
- Conformity marks: CE Approved
- Mounting: Horizontally only (bench or wall) using integral bracket
- Warranty: 1 year
Converter Tube:
- Length inside column oven: 10 in. (25.4 cm)
- Diameter: 1/2 in. (12.7 mm)
- Max. Inlet Pressure: 150 psi
- Max. Flow Rate: 1100 mL/min
signalword
Danger
hcodes
Hazard Classifications
Flam. Sol. 1
Storage Class
4.1B - Flammable solid hazardous materials
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service