Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

29542-U

Supelco

High Capacity Gas Purifier, Starter Kit

AC/DC input 110 V AC, size 1/4 in.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

size

1/4 in.

Looking for similar products? Visit Product Comparison Guide

General description

No other purifier removes both moisture and oxygen in such large quantities (35 liters of moisture, 14 liters of oxygen ).

Protects GC columns and detectors from damage caused by moisture and oxygen in the carrier gas stream, even when present up to 100 ppm levels. The clam-shell oven heats a converter tube to 580°C, causing moisture and oxygen to irreversibly react with the reactant material. This chemical reaction prevents contaminants from returning to the gas stream, even when the material approaches saturation, or when the oven is cooled. Carbon monoxide and carbon dioxide are also removed.

It is recommended to install 4 ft. (1.2 m) of tubing downstream of the oven to allow heat to dissipate from the gas stream. This tubing can be coiled to reduce space.

The starter kit includes a clam-shell oven, two elements (installed), and a converter tube.

Note: Not for use with hydrogen gas.

Clam-Shell Oven:
  • Length: 14 1/2 in. (36.8 cm)
  • Height: 5 1/4 in. (13.3 cm)
  • Depth: 6 in. (15.2 cm)
  • Operating Temp.: 580°C
  • Power Consumption: 90 watt
  • Conformity marks: CE Approved
  • Mounting: Horizontally only (bench or wall) using integral bracket
  • Warranty: 1 year

Converter Tube:
  • Length inside column oven: 10 in. (25.4 cm)
  • Diameter: 1/2 in. (12.7 mm)
  • Max. Inlet Pressure: 150 psi
  • Max. Flow Rate: 1100 mL/min

pictograms

Flame

signalword

Danger

hcodes

Hazard Classifications

Flam. Sol. 1

Storage Class

4.1B - Flammable solid hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  2. Does the High Capacity Gas Purifier contain asbestos?

    1 answer
    1. The High Capacity Gas Purifier does not contain asbestos.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service