29498-U
Closures with pre-cut septa, pk of 100
size 9 mm, blue polypropylene, red PTFE/silicone, size × thickness × 1.0 mm
About This Item
Recommended Products
material
blue polypropylene
red PTFE/silicone
description
9mm cap short thread
feature
center hole
hardness
55 (Durometer: shore A)
packaging
pack of 100
size × thickness
× 1.0 mm
size
9 mm
Looking for similar products? Visit Product Comparison Guide
General description
The Y-form shape of the cut was chosen for several reasons. Since the PTFE layer is uncut, this product provides protection against contamination until the needle pierces through it. The needle can easily penetrate through the Y-slit of the septa material (only the thickness of the PTFE layer needs to be penetrated by the needle). Less pressure is required for the autosampler needle to penetrate the pre-cut septa, resulting in fewer bent and/or broken needles.
The uncut PTFE layer provides a better seal than other commercially solid slitted septa until the needle punctures the septa. The Y-shaped cut allows air to ener the vial eliminating vacuum formation. The design provides the needle with a larger target area for penetration even when the needle doesn′t come down in the center of the septa.
related product
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service