29123-U
Interlocked™ vial with fused glass insert, screw top
volume 0.3 mL, amber glass vial, thread for 10-425, pkg of 100 ea
About This Item
material
amber glass vial
description
Interlocked vial (with fused-in insert)
feature
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 6 mm
volume
0.3 mL
fitting
thread for 10-425
Looking for similar products? Visit Product Comparison Guide
General description
- The flange top centers the inserts into the vial
- Glass to glass fusion eliminates concerns about contaminants
- Vials are made of borosilicate, Type 1 glass with a choice of clear or amber glass
Legal Information
related product
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service