Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28948-U

Supelco

Chromosorb® 106

stainless steel TD tube, unconditioned

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008

material

stainless steel TD tube

agency

ASTM® D6196

feature

unconditioned

packaging

pkg of 10 ea

O.D. × L

1/4 in. × 3 1/2 in.

matrix

Chromosorb 106

application(s)

industrial hygiene

compatibility

for use with PerkinElmer ATD-50,ATD-400 and TurboMatrix (TMX) Series

Looking for similar products? Visit Product Comparison Guide

Legal Information

ASTM is a registered trademark of American Society for Testing and Materials
Chromosorb is a registered trademark of Imerys Minerals California, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service