Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28863-U

Supelco

Super Clean (In-Line Design) Gas Purifier

hydrocarbon trap, stainless steel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
SB.54

material

stainless steel

Looking for similar products? Visit Product Comparison Guide

General description

Installation and replacement of the Click-On purifier is simple and eliminates the risk of damage to the gas line connections caused by overtightening. Simply attach the connectors to the gas line, install the purifier, and hand tighten the connectors.

Note: Does not include Click-on Connectors, these must be purchased before initial use.

Dimensions:
  • Cartridge and Connectors: 11 1/2 in. long x 1 1/4 in. diameter
  • Cartridge Only: 7 5/8 in. long x 1 1/4 in. diameter

pictograms

Health hazardEnvironment

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Resp. Sens. 1 - Skin Sens. 1 - STOT RE 2 Inhalation

target_organs

Lungs

Storage Class

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service