Key Documents
2879725-U
Supelco
Inlet Liner, Split/Splitless Type, Recessed Gooseneck Design (wool packed)
pkg of 25 ea
About This Item
UNSPSC Code:
41115719
NACRES:
SB.54
packaging
pkg of 25 ea
L × O.D. × I.D.
78.5 mm × 6.3 mm × 4.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Agilent (5890, 6890, and 7890)
  • Fits Finnigan (9001GCQ)
  • Fits Varian (CP-1177 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)