2878401-U
Inlet Liner, Split/Splitless Type, Single Taper FocusLiner™ Design (wool packed)
pkg of 1 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41115719
NACRES:
SB.54
description
Split/Splitless FocusLiner™ for 17A with SP-17, with taper (wool packed)
packaging
pkg of 1 ea
L × O.D. × I.D.
95 mm × 5.0 mm × 3.4 mm
Looking for similar products? Visit Product Comparison Guide
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
- Fits Shimadzu (17A) [with SPL-17 Injector]
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service