Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

2877305-U

Supelco

Inlet Liner, Splitless Type, Single Taper Design (unpacked)

pkg of 5 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115719
NACRES:
SB.54

description

Splitless for ThermoQuest 8000 and TRACE with taper

packaging

pkg of 5 ea

L × O.D. × I.D.

105 mm × 8.0 mm × 5.0 mm
105 mm × 8.0 mm × 5.0 mm

Looking for similar products? Visit Product Comparison Guide

General description

Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Thermo (ThermoQuest 8000 and TRACE)

Legal Information

TRACE is a trademark of Thermo Fisher Scientific or its subsidiaries

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service