Key Documents
2877201-U
Supelco
Inlet Liner, Splitless Type, Single Taper FocusLiner Design (wool packed)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
description
Splitless FocusLiner for ThermoQuest 8000 and TRACE (for use with 70mm needle)
packaging
pkg of 1 ea
L × O.D. × I.D.
105 mm × 8.0 mm × 5.0 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Thermo (ThermoQuest 8000 and TRACE)
Legal Information
FocusLiner is a trademark of SGE Analytical Science Pty Ltd.
TRACE is a trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)