Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28739-U

Supelco

Fused Silica Restrictor

for use with Agilent/HP 5973, 5890, 6890

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

compatibility

for use with Agilent/HP 5973, 5890, 6890

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service