Key Documents
28360-U
Supelco
domnick hunter® High-Purity Nitrogen Generator
Model 0751, 230 volt, 50 Hz, output flow, 750 cc/min
About This Item
UNSPSC Code:
41112500
NACRES:
SB.54
feature
output flow 750 cc/min
manufacturer/tradename
Domnick Hunter G1-110E
General description
These Nitrox nitrogen generators employ pressure swing adsorption technology to produce a continuous supply of high purity nitrogen. An integral oil-free air compressor delivers filtered compressed air (7 bar/101 psig) to a bed of carbon molecular sieve, which selectively removes oxygen, water vapor, carbon dioxide, and hydrocarbons. The adsorbent bed alternates between purification and regeneration modes, ensuring product nitrogen meets specified purity levels. A storage reservoir ensures a continuous supply of nitrogen. For units without an air compressor, please inquire.
Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at [email protected] or phone 800-359-3041.
Legal Information
domnick hunter is a registered trademark of Parker Hannifin
Choose from one of the most recent versions:
Certificates of Analysis (COA)