Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28358-U

Supelco

domnick hunter® Zero Air Generator

Model 10, 230 volt, output flow, 1000 cc/min

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500

feature

output flow 1000 cc/min

manufacturer/tradename

UHP-10ZA 230V
Domnick Hunter

Looking for similar products? Visit Product Comparison Guide

General description

  • Low power usage
  • Easy to wall mount
  • CE approved

Nitrox Zero Air Generators employ a catalytic oxidation system to produce a continuous supply of air that is virtually free of hydrocarbons, including methane. After incoming compressed air is filtered to remove oil, water, and particles, it is passed over a heated platinum catalyst that converts methane and oxygen in the air to carbon dioxide and water. If the hydrocarbon content of the input air is less than 10 ppm (as methane), the residual hydrocarbon concentration (as methane) is less than 0.1 ppm. A high efficiency 0.01 micron filter downstream of the catalyst ensures further removal of particles from the product stream.

Maintenance filter kits and warranty infomation available upon request. Contact Supelco Technical Service at [email protected] or phone 800-359-3041.

Legal Information

domnick hunter is a registered trademark of Parker Hannifin

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service