28349INT
Jumper Chip, Intuvo, Multimode Inlet
Synonym(s):
Intuvo Guard Chips and Jumper Chips
About This Item
manufacturer/tradename
Agilent Intuvo™
compatibility
for use with Agilent Intuvo GC
Looking for similar products? Visit Product Comparison Guide
General description
The Intuvo Guard Chip is a disposable consumable. To prolong the lifetime of your Intuvo GC column, replace the guard chip at the same frequency as you would typically trim a conventional GC column, or whenever chromatographic performance begins to decline. Replacement is fast, consistent, and simple, minimizing disruption to lab efficiency.
For some ultra-clean applications where a guard chip may not be needed, an Intuvo Jumper Chip provides a short, direct flow path. Customers whose typical applications do not require column trimming may be able to use jumper chips. Most customer applications will utilize guard chips rather than jumper ships to ensure column longevity.
Features and Benefits
- Saves time and improves sample throughput; no more column trimming
- Enables retention time reproducibility, and easy, consistent installation
- Excellent for use with GC/MS since there′s no need to vent the MS when replacing the guard chip
- Not reusable, replace regularly for best results
Jumper Chip
- Best suited for ultra-clean applications, especially those using headspace or purge and trap sample introduction techniques
- Direct path from inlet to column
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service