Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28347INT

Supelco

Guard Chip, Intuvo, Multimode Inlet

pk of 2 ea

Synonym(s):

Intuvo Guard Chips and Jumper Chips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115710

packaging

pk of 2 ea

manufacturer/tradename

Agilent Intuvo

compatibility

for use with Agilent Intuvo GC

Looking for similar products? Visit Product Comparison Guide

General description

An Agilent Intuvo Guard Chip saves time and improves sample throughput by eliminating the need for column trimming. The power of a guard column is harnessed in an innovative metal microfluidic chip. It traps sample residue by providing almost one meter of inert flow path between the inlet liner and the column. With regular replacement, sample contamination never reaches the head of the Intuvo GC column - eliminating the need for column trimming.
The Intuvo Guard Chip is a disposable consumable. To prolong the lifetime of your Intuvo GC column, replace the guard chip at the same frequency as you would typically trim a conventional GC column, or whenever chromatographic performance begins to decline. Replacement is fast, consistent, and simple, minimizing disruption to lab efficiency.

For some ultra-clean applications where a guard chip may not be needed, an Intuvo Jumper Chip provides a short, direct flow path. Customers whose typical applications do not require column trimming may be able to use jumper chips. Most customer applications will utilize guard chips rather than jumper ships to ensure column longevity.

Features and Benefits

Guard Chip
  • Saves time and improves sample throughput; no more column trimming
  • Enables retention time reproducibility, and easy, consistent installation
  • Excellent for use with GC/MS since there′s no need to vent the MS when replacing the guard chip
  • Not reusable, replace regularly for best results

Jumper Chip
  • Best suited for ultra-clean applications, especially those using headspace or purge and trap sample introduction techniques
  • Direct path from inlet to column

Legal Information

Intuvo is a trademark of Agilent Technologies, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service