Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28341-U

Supelco

SLB®-PAHms Capillary GC Column

L × I.D. 30 m × 0.25 mm, df 0.10 μm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
SB.54

material

fused silica

manufacturer/tradename

SLB®

parameter

25-350 °C temperature (isothermal)
25-360 °C temperature (programmed)

Beta value

625

df

0.10 μm

technique(s)

GC/MS: suitable
gas chromatography (GC): suitable

L × I.D.

30 m × 0.25 mm

application(s)

food and beverages

column type

capillary polar

Looking for similar products? Visit Product Comparison Guide

General description

Application: This special purpose and specially tested capillary GC column is designed for the analysis of polycyclic aromatic hydrocarbons (PAHs). It incorporates a high phenyl content polysiloxane stationary phase. Each column is individually tested to ensure resolution of several key sets (triphenylene/chrysene, and benzo[b]fluoranthene/benzo[k]fluoranthene/benzo[j]fluoranthene). The low bleed characteristics, inertness, and durable nature make it a great column anywhere a low bleed polar column is required.
USP Code: None
Phase: Bonded; Proprietary
Temp. Limits: ambient to 350 °C (isothermal) or 360 °C (programmed)

Legal Information

SLB is a registered trademark of Merck KGaA, Darmstadt, Germany

signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service