Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28188-U

Supelco

Detector Tube Pump

Toximeter II Automatic

Synonym(s):

TOXIMETER I

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151816
NACRES:
NB.24

application(s)

industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Kwik-Draw Deluxe Detector Tube Pump
  • One-hand operation - squeeze handle to compress bellows
  • Stroke counter
  • End-of-stroke indicator
  • Consistent 100mL sample draw

Gas-Tester II Detector Tube Pump
  • Stroke counter
  • End-of-stroke indicator
  • Unique Locking System
  • 100mL sample draw
  • Preset number of (100mL) pump strokes
  • Doubles as a sampling pump for other tubes - bags - impingers
  • Constant flow rate of 300cc/min for adsorbent tubes
  • 8 hours operating time
  • Fault recognition for low battery or blocked flow
  • Rechargeable NiCd battery pack included

Quality

CE approved.
Protection class: IP54; Ex Protection: EEx ia IIC T4, EEx ia I intrinsic safety approval. Other approvals pending.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service