Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28186-U

Supelco

Detector Tube Pump

Gas-Tester II H

Synonym(s):

GAS TESTER II H PUM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711

application(s)

industrial hygiene

Looking for similar products? Visit Product Comparison Guide

General description

Kwik-Draw Deluxe Detector Tube Pump
  • One-hand operation - squeeze handle to compress bellows
  • Stroke counter
  • End-of-stroke indicator
  • Consistent 100mL sample draw

Gas-Tester II Detector Tube Pump
  • Stroke counter
  • End-of-stroke indicator
  • Unique Locking System
  • 100mL sample draw
  • Stroke counter
  • End-of-stroke indicator
  • Unique locking system
  • 100mL sample draw

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service