Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

28155-U

Supelco

Omega Battery Charger

output12 V (adapter)

Synonym(s):

12 VOLT OMEGA BATTERY CHARGER

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104008
NACRES:
NB.24

technique(s)

active air sampling: suitable

output

12 V (adapter)

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with Escort Elf Air Sampling Pump

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service