Key Documents
27769-U
Desiccant Cartridge
About This Item
EC Number:
UNSPSC Code:
41112500
NACRES:
SB.54
manufacturer/tradename
60-497-0412
Domnick Hunter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Questions
1–4 of 4 Questions  
  1. Does a color change occur as the material is spent when using Product No. 27769-U Dessicant Cartridge?
    1 answer
    1. When the cartridge is new, the desiccant will be a mixture of blue and white material.The material turns grey as it is spent.
      Helpful?
  2. How is Product No. 27769-U Dessicant Cartridge used?
    1 answer
    1. This is a replacement part for domnick hunter hydrogen generators.
      Helpful?
  3. What is the Department of Transportation shipping information for this product?
    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.
      Helpful?
  4. How long does Product No. 27769-U Dessicant Cartridge last?
    1 answer
    1. The life of the cartridge will depend on gas usage. The cartridge does not need replaced until the color change is observed at the bottom of the viewing window.
      Helpful?