27468-U
Vials, screw top with solid cap with aluminum liner, preassembled, pkg of 100
volume 2 mL, amber glass vial (standard opening - 4.6 mm), O.D. × H × I.D. 12 mm × 32 mm × 4.6 mm, thread for 8-425
Synonym(s):
PK100 2ML AMBER S/T VIAL, SOLID CAP W/ALUMINUM LINER
About This Item
Recommended Products
material
aluminum liner
amber glass vial (standard opening - 4.6 mm)
black phenolic solid cap
feature
closure type black phenolic solid top cap
closure type screw top vial
packaging
pkg of 100 ea
O.D. × H × I.D.
12 mm × 32 mm × 4.6 mm
volume
2 mL
fitting
thread for 8-425
Looking for similar products? Visit Product Comparison Guide
related product
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service