27388
Vials, screw top, clear glass
volume 10 mL, O.D. × H 24.5 mm × 52 mm, thread for 22-400
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121806
Recommended Products
material
clear glass vial
description
cap not included
feature
closure type screw top vial
packaging
pkg of × 144 ea
O.D. × H
24.5 mm × 52 mm
volume
10 mL
fitting
thread for 22-400
Looking for similar products? Visit Product Comparison Guide
General description
Vials are suitable basically to inject samples from an autosampler. Autosampler needle pierces through the cap during injection and withdraws the required aliquots of sample from the vials. Screw top vials (volume 10 mL, O.D. × H 24.5 mm × 52 mm) are used on many autosamplers. The threads on the vials are 22-400.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service