Key Documents
27321
Supelco
Shimadzu Autosampler Vials
5mL vial and closure, pre-assembled, O.D. × H 12 mm × 96 mm, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
feature
closure type black polypropylene hole cap
closure type screw top vial
pre-assembled
packaging
pkg of 100 ea
O.D. × H
12 mm × 96 mm
volume
5 mL
fitting
thread for 10-425
General description
This 5mL sample vial assembly designed for use in Shimadzu Model ASI-5000 for automated total organic carbon analysis. The vial assembly is designed to be used with a 10-425 threaded cap which allows the vials to become suspended in the 78 vial regular turntable. Part number 2-7321 includes the attached polypropylene cap.
Legal Information
Shimadzu is a trademark of Shimadzu Corporation
Choose from one of the most recent versions:
Certificates of Analysis (COA)