Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27271

Supelco

Screw cap (open-top), polypropylene

black polypropylene cap, for use with 2 mL vial (large opening), pkg of × 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

black polypropylene cap

feature

autoclavable
closure type black polypropylene hole cap

packaging

pkg of × 100 ea

parameter

135 °C temp. limit

diam. × H

13 mm × 9.5 mm

hole diam.

7 mm

fitting

thread for 10-425

compatibility

for use with 2 mL vial (large opening)

Looking for similar products? Visit Product Comparison Guide

General description

These hole caps do not come with septa.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula Moyano et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 129, 87-96 (2019-04-29)
Amitraz is a neurotoxic formamidine pesticide that induces cell death in hippocampal neurons, although its mechanisms are unknown. Amitraz produces reactive oxygen species (ROS), which could lead to cell death. Amitraz was shown to induce different cytochrome P450 (CYP) isoenzymes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service