Key Documents
27264-U
Supelco
Work Station, nine-drawer
filled with 2000 screw top vials and closures
, vial volume 2 mL, clear glass vial, kit of 1 ea
About This Item
UNSPSC Code:
41122001
material
black phenolic cap
clear glass vial
description
septum type: PTFE/silicone
feature
closure type black polypropylene hole cap
packaging
kit of 1 ea
vial volume
2 mL
General description
A convenient way to organize your vials. Available filled with supplies or unfilled. Cat. No. 27231-U includes 3000 clear large-opening 2 mL vials, PTFE/red rubber 11 mm septa, and seals. Cat. No. 27264-U includes 2000 clear screw top 2 mL vials (standard opening), red PTFE/silicone septa, and black phenolic closures (recommended for Varian and Spectra Physics autosamplers).
Choose from one of the most recent versions:
Certificates of Analysis (COA)