Key Documents
27107
Supelco
Crimp seals with Viton® septa
silver aluminum seal, seal diam. 11 mm, black Viton® septum, septum thickness 30 mil, pkg of 100 ea
About This Item
UNSPSC Code:
41122001
material
black Viton® septum
silver aluminum seal
feature
closure type crimp seal
closure type crimp seal
packaging
pkg of 100 ea
seal diam.
11 mm
septum thickness
30 mil
fitting
thread for 11 mm
Legal Information
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)