Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27104

Supelco

Crimp seals, 11 mm, with PTFE/rubber septa

blue aluminum seal, seal diam. 11 mm, PTFE/red rubber septum, pkg of 100 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121821

material

PTFE/red rubber septum
blue aluminum seal

packaging

pkg of 100 ea

O.D.

11 mm

seal diam.

11 mm

fitting

thread for 11 mm

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sunanda Margrett Williams et al.
Structure (London, England : 1993), 25(9), 1449-1454 (2017-08-22)
Proteins of the ferritin family are ubiquitous in living organisms. With their spherical cage-like structures they are the iron storehouses in cells. Subfamilies of ferritins include 24-meric ferritins and bacterioferritins (maxiferritins), and 12-meric Dps (miniferritins). Dps safeguards DNA by direct
Karolina Hus et al.
Frontiers in plant science, 11, 614-614 (2020-06-09)
The CRISPR/Cas9 system enables precise genome editing and is a useful tool for functional genomic studies. Here we report a detailed protocol for targeted genome editing in the model grass Brachypodium distachyon and its allotetraploid relative B. hybridum, describing gRNA

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service