Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

27035

Supelco

Micro-Reaction Vessel, black phenolic solid cap, pkg of 12

volume 0.3 mL, clear glass vial, styrene-butadiene septum, O.D. × H 16 mm × 32 mm

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001

material

clear glass vial
styrene-butadiene septum

feature

closure type black phenolic solid top cap
closure type screw top vial
graduations
pre-assembled

packaging

pkg of 12 ea

O.D. × H

16 mm × 32 mm

volume

0.3 mL

fitting

thread for 15-425

Looking for similar products? Visit Product Comparison Guide

Application

It has been used to check the reaction temperature and length during examining background fatty acids signals in GC/MS.[1] It was used in enzymatic assay, a modified spectrophotometric assay for Peptidyl dipeptidase determination in serum.[2]

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Micro-quantitation of lipids in serum-free cell culture media: a critical aspect is the minimization of interference from medium components and chemical reagents.
Shen, Chun Fang, Jalal Hawari, and Amine Kamen.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 810.1, 119-127 (2004)
A liquid chromatography-assisted assay for angiotensin-converting enzyme (peptidyl dipeptidase) in serum.
Chiknas, Steven G.
Clinical Chemistry, 25.7, 1259-1262 (1979)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service