Key Documents
26409
Supelco
Inlet Liner, Packed Column, for PerkinElmer® 8000, Sigma® 2000/1200, 1B-4B & 300
pkg of 5 ea
About This Item
UNSPSC Code:
41115700
packaging
pkg of 5 ea
compatibility
configured for models manufactured 1978 or later
General description
  • length 101 mm × O.D. 4.6 mm × I.D. 2.75 mm
  • original Mfr part no. 03302221
·
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Sigma is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)