2637805
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 5 ea
All Photos(1)
About This Item
UNSPSC Code:
41115719
Recommended Products
description
SPME/Splitless
packaging
pkg of 5 ea
L × O.D. × I.D.
54 mm × 5.0 mm × 0.8 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
- Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)