2637801
Inlet Liner, Direct (SPME) Type, Straight Design (unpacked)
pkg of 1 ea
Select a Size
Select a Size
About This Item
Recommended Products
description
SPME/Splitless
packaging
pkg of 1 ea
manufacturer/tradename
Varian 392611948
L × O.D. × I.D.
54 mm × 5.0 mm × 0.8 mm
compatibility
for use with Varian 1078 Injector
technique used for SPME
General description
- Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)