Key Documents
2637605
Supelco
Inlet Liner, Direct Type, Straight Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
41115719
description
Splitless/Trace analytes
packaging
pkg of 5 ea
L × O.D. × I.D.
54 mm × 5.0 mm × 0.5 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)