Key Documents
2637601
Supelco
Inlet Liner, Direct Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
description
Splitless/Trace analytes
packaging
pkg of 1 ea
manufacturer/tradename
Varian 03-925331-00 (OEM)
L × O.D. × I.D.
54 mm × 5.0 mm × 0.5 mm
compatibility
for use with Varian 1078 Injector
technique used for Trace analytes; Thermolabile and polar compounds
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)