Key Documents
2637205
Supelco
Inlet Liner, Split Type, Frit Design (unpacked)
pkg of 5 ea
About This Item
UNSPSC Code:
41115719
packaging
pkg of 5 ea
L × O.D. × I.D.
54 mm × 5.0 mm × 3.4 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)