Key Documents
2636325
Supelco
Inlet Liner, Direct Type, Straight Design (unpacked)
pkg of 25 ea
About This Item
UNSPSC Code:
12352200
description
Direct high performance
packaging
pkg of 25 ea
L × O.D. × I.D.
54 mm × 4.6 mm × 0.5 mm
54 mm × 4.6 mm × 0.5 mm
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1093-94 SPI Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)