Key Documents
2636201
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
12000000
material
fused silica
description
Splitless
packaging
pkg of 1 ea
L × O.D. × I.D.
72 mm × 6.3 mm × 4.0 mm
compatibility
for use with Varian 1075/1077
technique used for High linearity (0.25/0.32 mm I.D. columns)
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Other Notes
I263620 --> material nos. 2636201 (1pk) & 2636205 (5pk).
Choose from one of the most recent versions:
Certificates of Analysis (COA)