Key Documents
2636101
Supelco
Inlet Liner, Splitless Type, Straight Design (unpacked)
pkg of 1 ea
About This Item
UNSPSC Code:
12352200
description
Split (unpacked)
unpacked
packaging
pkg of 1 ea
manufacturer/tradename
Varian 16-000830-00 (OEM, instrument manufacturer′s nondeactivated liner)
L × O.D. × I.D.
72 mm × 6.3 mm × 4.0 mm
compatibility
configured for
for use with Varian 1075/1077 Injectors
technique used for High & low MW analytes
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1075 and 1077 Injector)
Other Notes
I263610 --> material nos. 2636101 (1pk), 2636105 (5pk), 2636125 (25pk).
Footnote
Instrument manufacturer′s nondeactivated liner.
Choose from one of the most recent versions:
Certificates of Analysis (COA)